missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10471553
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-95-3
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00005992
InChI Key XJLSEXAGTJCILF-UHFFFAOYSA-N
Synonym nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine
PubChem CID 4498
ChEBI CHEBI:116931
IUPAC Name piperidine-3-carboxylic acid
SMILES C1CC(CNC1)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 129.16
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 3-Piperidinecarboxylic acid, Functional Group amine (alkyl)
     carboxylic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'