missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidin-1-ylmethyl benzylamine, ≥97%, Thermo Scientific™

Product Code. 10081004
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10081004 5 g 5g
10513423 250 mg 250mg
10219932 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10081004 Supplier Thermo Scientific Maybridge Supplier No. MO01157DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 91271-80-6
Molecular Formula C13H20N2
Molecular Weight (g/mol) 204.317
MDL Number MFCD06408781
InChI Key BMNFPARGJHNMAX-UHFFFAOYSA-N
Synonym 3-piperidin-1-ylmethyl-benzylamine, 1-3-piperidin-1-ylmethyl phenyl methanamine, 3-piperidin-1-ylmethyl phenyl methanamine, 3-piperidin-1-ylmethyl benzyl amine, 3-piperidylmethyl phenyl methylamine, 3-piperidin-1-ylmethylbenzylamine, benzenemethanamine, 3-1-piperidinylmethyl, 3-piperidin-1-yl methyl phenyl methanamine, 1-3-piperidin-1-ylmethyl phenyl methylamine
PubChem CID 2794678
IUPAC Name [3-(piperidin-1-ylmethyl)phenyl]methanamine
SMILES C1CCN(CC1)CC2=CC(=CC=C2)CN

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 204.32
Percent Purity 97%
Chemical Name or Material 3-Piperidin-1-ylmethyl benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.