missing translation for 'onlineSavingsMsg'
Learn More

3-(Piperidin-1-ylmethyl)aniline, ≥97%, Thermo Scientific™

Product Code. 10312115
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10312115 5 g 5g
10538053 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10312115 Supplier Thermo Scientific Maybridge Supplier No. CC45214DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 93138-55-7
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD04114502
InChI Key SEJNYLBEEMVJNN-UHFFFAOYSA-N
Synonym 3-piperidin-1-ylmethyl aniline, 3-piperidin-1-ylmethyl phenyl amine, 3-1-piperidylmethyl aniline, 3-piperidylmethyl phenylamine, 3-1-piperidinylmethyl aniline, 3-piperidylmethyl aniline, benzenamine, 3-1-piperidinylmethyl, 3-piperidinylmethyl aniline, 1-3-aminobenzyl piperidine
PubChem CID 6484331
IUPAC Name 3-(piperidin-1-ylmethyl)aniline
SMILES C1CCN(CC1)CC2=CC(=CC=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 190.29
Percent Purity 97%
Chemical Name or Material 3-(Piperidin-1-ylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.