missing translation for 'onlineSavingsMsg'
Learn More

3-(Piperazin-1-yl)-1,2-benzisoxazole, 95%, Thermo Scientific™

Product Code. 12095400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12095400 5 g 5g
12085400 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12095400 Supplier Thermo Scientific Acros Supplier No. 446700050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 87691-89-2
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.24
InChI Key ZDFQBFVFCPABKQ-UHFFFAOYSA-N
Synonym 3-piperazin-1-yl benzo d isoxazole, 3-1-piperazinyl-1,2-benzisoxazole, 3-piperazin-1-yl-1,2-benzoxazole, 3-piperazin-1-yl-1,2-benzisoxazole, 3-1-piperazino-1,2-benzisoxazole, 3-piperazinylbenzo d isoxazole, 3-piperazin-1-yl-benzo d isoxazole, 3-1-piperazinyl-1,2-benzoxazole, 4 1,2-benzisoxazol-3-yl piperazine
PubChem CID 13232475
IUPAC Name 3-piperazin-1-yl-1,2-benzoxazole
SMILES C1CN(CCN1)C2=NOC3=CC=CC=C32

Specifications

Melting Point 69.0°C to 72.0°C
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material 3-(Piperazin-1-yl)-1, 2-benzisoxazole
compliance-icons
Product Identifier
  • 3-(Piperazin-1-yl)-1,2-benzisoxazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.