missing translation for 'onlineSavingsMsg'
Learn More

3-(Piperazin-1-yl)-1,2-benzisoxazole, 95%, Thermo Scientific™

Product Code. 12085400
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 87691-89-2
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.24
InChI Key ZDFQBFVFCPABKQ-UHFFFAOYSA-N
Synonym 3-piperazin-1-yl benzo d isoxazole, 3-1-piperazinyl-1,2-benzisoxazole, 3-piperazin-1-yl-1,2-benzoxazole, 3-piperazin-1-yl-1,2-benzisoxazole, 3-1-piperazino-1,2-benzisoxazole, 3-piperazinylbenzo d isoxazole, 3-piperazin-1-yl-benzo d isoxazole, 3-1-piperazinyl-1,2-benzoxazole, 4 1,2-benzisoxazol-3-yl piperazine
PubChem CID 13232475
IUPAC Name 3-piperazin-1-yl-1,2-benzoxazole
SMILES C1CN(CCN1)C2=NOC3=CC=CC=C32

Specifications

Melting Point 69.0°C to 72.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material 3-(Piperazin-1-yl)-1, 2-benzisoxazole
compliance-icons
Product Identifier
  • 3-(Piperazin-1-yl)-1,2-benzisoxazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'