missing translation for 'onlineSavingsMsg'
Learn More

3-Picolyl chloride hydrochloride, 99%

Product Code. 10269503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269503 500 g Glass Bottle 500g
10026253 5 g Glass Bottle 5g
10152990 25 g Glass Bottle 25g
10785601 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10269503 Supplier Thermo Scientific Acros Supplier No. 131195000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6959-48-4 | C6H6ClN·HCl | 164.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6959-48-4
Molecular Formula C6H6ClN·HCl
Molecular Weight (g/mol) 164.04
MDL Number MFCD00012818
InChI Key UZGLOGCJCWBBIV-UHFFFAOYSA-N
Synonym 3-chloromethyl pyridine hydrochloride, 3-chloromethylpyridine hydrochloride, 3-picolyl chloride hydrochloride, 3-chloromethyl pyridine hcl, ccris 140, pyridine, 3-chloromethyl-, hydrochloride, 3-pyridylmethyl chloride hydrochloride, unii-bqa8fyp4yn, 3-chloromethyl pyridinium chloride, 3-chloromethyl pyridine.hcl
PubChem CID 23394
IUPAC Name 3-(chloromethyl)pyridine;hydrochloride
SMILES C1=CC(=CN=C1)CCl.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 137°C to 143°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 500 g
Solubility Information Solubility in water: 820g/L
Formula Weight 164.04
Percent Purity 99%
Physical Form Crystalline Needles or Powder
Chemical Name or Material 3-Picolyl chloride hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.