missing translation for 'onlineSavingsMsg'
Learn More

3-Phthalimidopropionic acid, 98%

Product Code. 11353127
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353127 1 g 1g
11363127 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353127 Supplier Thermo Scientific Alfa Aesar Supplier No. L13535.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3339-73-9 | C11H9NO4 | 219.196 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3339-73-9
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
MDL Number MFCD00023096
InChI Key DXXHRZUOTPMGEH-UHFFFAOYSA-N
Synonym 3-phthalimidopropionic acid, 3-1,3-dioxoisoindolin-2-yl propanoic acid, 2h-isoindole-2-propanoic acid, 1,3-dihydro-1,3-dioxo, 3-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 3-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, pht-beta-ala-oh, b-phthalimidopropionic acid, 3-phthalimidopropanoic acid, n-phthaloyl-beta-alanine, phthalyl-.beta.-alanine
PubChem CID 76859
IUPAC Name 3-(1,3-dioxoisoindol-2-yl)propanoic acid
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCC(=O)O

Specifications

Melting Point 151°C to 153°C
Quantity 1 g
Beilstein 15570
Formula Weight 219.2
Percent Purity 98%
Chemical Name or Material 3-Phthalimidopropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.