missing translation for 'onlineSavingsMsg'
Learn More

3-Phthalimidopropanoyl Chloride (Technical Grade), TRC

Product Code. 30373001
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373001 250 mg
30372666 100 mg
30379834 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30373001 Supplier Toronto Research Chemicals Supplier No. TRCD462610250MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17137-11-0
Molecular Formula C11H8ClNO3
Molecular Weight (g/mol) 237.64
Synonym 3-Phthalimidopropionyl Chloride, 3-Phthalimidoylpropionyl Chloride, Phthaloyl-beta-alanyl Chloride, beta-Phthalimidopropionic Acid Chloride, 3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propionyl Chloride, 3-(1,3-Dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoyl Chloride
IUPAC Name 3-(1,3-dioxoisoindol-2-yl)propanoyl chloride
SMILES ClC(=O)CCN1C(=O)c2ccccc2C1=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H8ClNO3/c12-9(14)5-6-13-10(15)7-3-1-2-4-8(7)11(13)16/h1-4H,5-6H2
Formula Weight 237.0193
Chemical Name or Material 3-Phthalimidopropanoyl Chloride (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.