missing translation for 'onlineSavingsMsg'
Learn More

(3-Phenylpropyl)triphenylphosphonium bromide, 97+%

Product Code. 11460543
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460543 50 g 50g
11450543 10 g 10g
11470543 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460543 Supplier Thermo Scientific Alfa Aesar Supplier No. A12669.18
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7484-37-9 | C27H26BrP | 461.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7484-37-9
Molecular Formula C27H26BrP
Molecular Weight (g/mol) 461.38
MDL Number MFCD00051886
InChI Key RPUZOJFXAPSSJD-UHFFFAOYSA-M
Synonym 3-phenylpropyl triphenylphosphonium bromide, triphenyl 3-phenylpropyl phosphonium bromide, triphenyl 3-phenylpropyl phosphanium bromide, 3-phenylpropyltriphenylphosphoniumbromide, phenylpropyl triphenylphosphonium bromide, 3-phenylpropyltriphenylphosphonium bromide, triphenyl 3-phenylpropylphosphonium bromide, 3-phenyl-propyl-triphenylphosphonium bromide, 3-phenylpropyl triphenyl phosphonium bromide
PubChem CID 10928647
IUPAC Name triphenyl(3-phenylpropyl)phosphanium;bromide
SMILES [Br-].C(CC1=CC=CC=C1)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 208°C to 212°C
Odor Odorless
Linear Formula C6H5(CH2)2P(C6H5)3Br
Quantity 50 g
Beilstein 4173336
Sensitivity Hygroscopic
Formula Weight 461.39
Percent Purity 98%
Chemical Name or Material (3-Phenylpropyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.