missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionyl chloride, 98%

Product Code. 11469842
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469842 250 g 250g
11449842 10 g 10g
11459842 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469842 Supplier Thermo Scientific Alfa Aesar Supplier No. A10860.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 645-45-4 | C9H9ClO | 168.62 g/mol

The 3-phenylpropionate (dihydrocinnamoyl) ester has been used as a protecting group in nucleoside synthesis. Used in the synthesis of 1-Phenyl-3-methyl-4-acyl-5- pyrazolones. N-(3-phenyl-n-propyl)-1-phenyl-2-aminopropane were prepared by acylation of the appropriate amine with 3-phenylpropionyl chloride followed by reduction of the resulting amide with either LiA1H4. For the preparation of the phenylacetyl (PA) and 3-phenylpropionyl (PP) derivatives, the corresponding amino acids were coupled in the usual manner with phenylacetyl chloride or 3-phenylpropionyl chloride in the presence of either NaOH(PA-LD- and DL-Pgly. 3-Phenylpropionyl chloride (4) was treated with diazomethane to give diazoketone. Allyloxy resin 12 was acylated with 3-phenylpropionyl chloride and treated with 50% TFA for 1 h followed by treatment with Pd(PPh3)4/PPh3 in solution to furnish the desired hydrox- amic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The 3-phenylpropionate (dihydrocinnamoyl) ester has been used as a protecting group in nucleoside synthesis. Used in the synthesis of 1-Phenyl-3-methyl-4-acyl-5- pyrazolones. N-(3-phenyl-n-propyl)-1-phenyl-2-aminopropane were prepared by acylation of the appropriate amine with 3-phenylpropionyl chloride followed by reduction of the resulting amide with either LiA1H4. For the preparation of the phenylacetyl (PA) and 3-phenylpropionyl (PP) derivatives, the corresponding amino acids were coupled in the usual manner with phenylacetyl chloride or 3-phenylpropionyl chloride in the presence of either NaOH(PA-LD- and DL-Pgly. 3-Phenylpropionyl chloride (4) was treated with diazomethane to give diazoketone. Allyloxy resin 12 was acylated with 3-phenylpropionyl chloride and treated with 50% TFA for 1 h followed by treatment with Pd(PPh3)4/PPh3 in solution to furnish the desired hydrox- amic acid.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, amines, alcohols, active metals, bases, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-45-4
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00000748
InChI Key MFEILWXBDBCWKF-UHFFFAOYSA-N
Synonym 3-phenylpropionyl chloride, hydrocinnamoyl chloride, benzenepropanoyl chloride, dihydrocinnamoyl chloride, hydrocinnamyl chloride, propionyl chloride, 3-phenyl, hydrocinnamoylchloride, beta-phenylpropionyl chloride, 3-phenylpropionic acid chloride, beta-phenylpropanoyl chloride
PubChem CID 64801
IUPAC Name 3-phenylpropanoyl chloride
SMILES C1=CC=C(C=C1)CCC(=O)Cl

Specifications

Melting Point -7°C
Density 1.135
Boiling Point 225°C
Flash Point 108°C (226°F)
Odor Acrid
Refractive Index 1.5265
Quantity 250 g
UN Number UN3265
Beilstein 742586
Sensitivity Moisture sensitive
Solubility Information Soluble in water (reacts),and ether.
Formula Weight 168.62
Percent Purity 98%
Chemical Name or Material 3-Phenylpropionyl chloride
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.