missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionaldehyde, 95%

Product Code. 11474173
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474173 50 g 50g
11484173 250 g 250g
11464173 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474173 Supplier Thermo Scientific Alfa Aesar Supplier No. A10367.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-53-0 | C9H10O | 134.178 g/mol

3-Phenylpropionaldehyde is used as a flavoring agent. It is also used in the preparation of cinnamaldehyde, 2-chloro hydrocinnamaldehyde by alfa chlorination and mixture of homopropargyl alcohols. It serves as a precursor for the synthesis of mixture of syn- and anti-beta-hydroxyallylsilanes by hydroboration of allenylsilane. It is also involved in the catalytic asymmetric cyanosilylation to prepare (2S)-2-hydroxy-4-phenylbutanenitrile. Further, it undergoes Henry reaction to form nitroaldols by reacting with nitromethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpropionaldehyde is used as a flavoring agent. It is also used in the preparation of cinnamaldehyde, 2-chloro hydrocinnamaldehyde by alfa chlorination and mixture of homopropargyl alcohols. It serves as a precursor for the synthesis of mixture of syn- and anti-beta-hydroxyallylsilanes by hydroboration of allenylsilane. It is also involved in the catalytic asymmetric cyanosilylation to prepare (2S)-2-hydroxy-4-phenylbutanenitrile. Further, it undergoes Henry reaction to form nitroaldols by reacting with nitromethane.

Solubility
Miscible with chloroform, dichloromethane, ethyl acetate, alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-53-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00007021
InChI Key YGCZTXZTJXYWCO-UHFFFAOYSA-N
Synonym benzenepropanal, 3-phenylpropionaldehyde, hydrocinnamaldehyde, hydrocinnamic aldehyde, dihydrocinnamaldehyde, benzylacetaldehyde, 3-phenyl-1-propanal, 3-phenylpropylaldehyde, 3-phenylpropyl aldehyde, propanal, phenyl
PubChem CID 7707
IUPAC Name 3-phenylpropanal
SMILES C1=CC=C(C=C1)CCC=O

Specifications

Melting Point °C to 42°C
Density 1.017
Boiling Point 222°C to 223°C
Flash Point 95°C (203°F)
Linear Formula C6H5(CH2)2CHO
Refractive Index 1.523
Quantity 50 g
Beilstein 1071910
Sensitivity Air Sensitive
Solubility Information Miscible with chloroform,dichloromethane,ethyl acetate,alcohol and ether. Immiscible with water.
Formula Weight 134.18
Percent Purity 95%
Chemical Name or Material 3-Phenylpropionaldehyde
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.