missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionaldehyde, 94.9%, MP Biomedicals™

Product Code. 11413562 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413562 100 g 100g
11403562 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413562 Supplier MP Biomedicals Supplier No. 0215619680
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Tilføj til indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolitik

Kemiske identifikatorer

CAS 104-53-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
InChI Key YGCZTXZTJXYWCO-UHFFFAOYSA-N
Synonym benzenepropanal, 3-phenylpropionaldehyde, hydrocinnamaldehyde, hydrocinnamic aldehyde, dihydrocinnamaldehyde, benzylacetaldehyde, 3-phenyl-1-propanal, 3-phenylpropylaldehyde, 3-phenylpropyl aldehyde, propanal, phenyl
PubChem CID 7707
IUPAC Name 3-phenylpropanal
SMILES C1=CC=C(C=C1)CCC=O

Tekniske data

Color Colorless
Density 1.01g/mL
Assay Percent Range 95%
Quantity 100 g
Formula Weight 134.2
Percent Purity 0.949
Physical Form Liquid
Chemical Name or Material 3-Phenylpropionaldehyde
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.