missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionaldehyde, 94.9%, MP Biomedicals™

Product Code. 11403562 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403562 25 g 25g
11413562 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403562 Supplier MP Biomedicals Supplier No. 0215619625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Identificadores de productos químicos

CAS 104-53-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
InChI Key YGCZTXZTJXYWCO-UHFFFAOYSA-N
Synonym benzenepropanal, 3-phenylpropionaldehyde, hydrocinnamaldehyde, hydrocinnamic aldehyde, dihydrocinnamaldehyde, benzylacetaldehyde, 3-phenyl-1-propanal, 3-phenylpropylaldehyde, 3-phenylpropyl aldehyde, propanal, phenyl
PubChem CID 7707
IUPAC Name 3-phenylpropanal
SMILES C1=CC=C(C=C1)CCC=O

Especificaciones

Color Colorless
Density 1.01g/mL
Assay Percent Range 95%
Quantity 25 g
Formula Weight 134.2
Percent Purity 0.949
Physical Form Liquid
Chemical Name or Material 3-Phenylpropionaldehyde
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.