missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylisoxazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10566514
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 72418-40-7
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD09817543
InChI Key SZXMDUKWIYIKKS-UHFFFAOYSA-N
Synonym 3-phenylisoxazole-5-carbaldehyde, 3-phenyl-5-isoxazolecarbaldehyde, acmc-20agcz, 3-phenylisoxazole-5-aldehyde, 3-phenyl-isoxazole-5-aldehyde, 3-phenylisoxazole-5-carboxaldehyde, 5-isoxazolecarboxaldehyde,3-phenyl
PubChem CID 11367267
IUPAC Name 3-phenyl-1,2-oxazole-5-carbaldehyde
SMILES C1=CC=C(C=C1)C2=NOC(=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 173.17
Percent Purity 97%
Chemical Name or Material 3-Phenylisoxazole-5-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'