missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenylaminomethyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. p-7030378
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337649 1 g 1g
11347649 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337649 Supplier Thermo Scientific Alfa Aesar Supplier No. H28287.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073353-90-8
Molecular Formula C19H24BNO2
Molecular Weight (g/mol) 309.216
MDL Number MFCD09266198
InChI Key FMCOCBRSLHQLQR-UHFFFAOYSA-N
Synonym 3-phenylaminomethyl benzeneboronic acid pinacol ester, n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl aniline, n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl aniline, amtb061, 3-phenylaminomethyl phenylboronic acid pinacol ester, 3-phenylaminomethyl phenyl boronic acid pinacol ester, 3-phenylamino methyl phenylboronic acid, pinacol ester, 2-3-anilinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl benzenamine
PubChem CID 46739047
IUPAC Name N-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CNC3=CC=CC=C3

Specifications

Melting Point 112°C to 115°C
Quantity 1 g
Formula Weight 309.21
Percent Purity 97%
Chemical Name or Material 3-(Phenylaminomethyl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.