missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylacrylaldehyde oxime, 97%, Thermo Scientific™

Product Code. 10007102
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10007102 10 g 10g
10649404 1 g 1g
10624375 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10007102 Supplier Thermo Scientific Maybridge Supplier No. BTB09548EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 13372-81-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00019969
InChI Key RUQDOYIAKHIMAN-DAAQNPAKSA-N
Synonym cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z
PubChem CID 9561350
IUPAC Name (NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine
SMILES O\N=C\C=C\C1=CC=CC=C1

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 147.18
Chemical Name or Material 3-Phenylacrylaldehyde oxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.