missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-5-vinyl-1,2,4-oxadiazole, TRC

Product Code. 30012063
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012063 10 mg 10mg
30003557 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012063 Supplier Toronto Research Chemicals Supplier No. TRCP33832010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28917-17-1
Molecular Formula C10 H8 N2 O
Molecular Weight (g/mol) 172.183
Synonym 5-Ethenyl-3-phenyl-1,2,4-oxadiazole
IUPAC Name 3-phenyl-5-vinyl-1,2,4-oxadiazole
SMILES C=Cc1onc(n1)c2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H8N2O/c1-2-9-11-10(12-13-9)8-6-4-3-5-7-8/h2-7H,1H2
Formula Weight 172.0637
Percent Purity >95%
Chemical Name or Material 3-Phenyl-5-vinyl-1,2,4-oxadiazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.