missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1H-pyrazole-4-carboxaldehyde, 98%, Thermo Scientific™

Product Code. 10492831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10492831 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10492831 Supplier Thermo Scientific Acros Supplier No. 296000010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 26033-20-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 146°C to 148°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C10H8N2O
MDL Number MFCD00216971
Quantity 1 g
Synonym 3-phenyl-1h-pyrazole-4-carbaldehyde, 3-phenyl-1h-pyrazole-4-carboxaldehyde, 3-phenylpyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-phenyl, 3-phenyl-2h-pyrazole-4-carbaldehyde, 5-phenylpyrazole-4-carbaldehyde, pubchem16274, acmc-1cd6r, 3-phenyl-4-formylpyrazole, cambridge id 5132647
Solubility Information Solubility in water: 25mg/mL (100°C). Other solubilities: 400mg/mL ethanol (78°C)
InChI Key OCCFXKQCKSLEII-UHFFFAOYSA-N
SMILES O=CC1=C(NN=C1)C1=CC=CC=C1
IUPAC Name 5-phenyl-1H-pyrazole-4-carbaldehyde
Molecular Weight (g/mol) 172.19
PubChem CID 291816
Formula Weight 172.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Phenyl-1H-pyrazole-4-carboxaldehyde, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Phenyl-1H-pyrazole-4-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.