missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propylamine, 98%

Product Code. 10765601 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10765601 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10765601 Supplier Thermo Scientific Acros Supplier No. 130880250
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2038-57-5 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2038-57-5
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to White
Density 0.9500g/mL
Boiling Point 221°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Molecular Formula C9H13N
Linear Formula C6H5(CH2)3NH2
Refractive Index 1.5240 to 1.526
MDL Number MFCD00008224
Quantity 25 g
Beilstein 12, 1145
Specific Gravity 0.95
Synonym 3-phenylpropylamine, benzenepropanamine, 1-amino-3-phenylpropane, hydrocinnamylamine, gamma-phenylpropylamine, 3-phenyl-n-propylamine, 3-phenyl-1-propylamine, 3-phenyl-1-propanamine, propylamine, 3-phenyl, 3-phenylpropanamine
InChI Key LYUQWQRTDLVQGA-UHFFFAOYSA-N
SMILES NCCCC1=CC=CC=C1
IUPAC Name 3-phenylpropan-1-amine
Molecular Weight (g/mol) 135.21
PubChem CID 16259
Formula Weight 135.21
Percent Purity 98%
Physical Form Crystals or Flakes
Chemical Name or Material 3-Phenyl-1-propylamine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Phenyl-1-propylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.