missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propyl acetate, 98%

Product Code. 11449906
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449906 250 g 250g
11429906 10 g 10g
11439906 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449906 Supplier Thermo Scientific Alfa Aesar Supplier No. B20878.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 122-72-5 | C11H14O2 | 178.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-72-5
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00026216
InChI Key JRJGKUTZNBZHNK-UHFFFAOYSA-N
Synonym hydrocinnamyl acetate, benzenepropanol, acetate, phenylpropyl acetate, 3-phenyl-1-propyl acetate, 3-acetoxypropyl benzene, 3-phenyl-1-propanol, acetate, 3-acetoxy-1-phenylpropane, 1-acetoxy-3-phenylpropane, benzenepropanol, 1-acetate, benzenepropyl acetate
PubChem CID 31226
ChEBI CHEBI:81257
IUPAC Name 3-phenylpropyl acetate
SMILES CC(=O)OCCCC1=CC=CC=C1

Specifications

Density 1.01
Boiling Point 252°C
Flash Point 130°C (266°F)
Refractive Index 1.496
Quantity 250 g
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material 3-Phenyl-1-propyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.