missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxytoluene, 97%

Product Code. 11484967
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484967 25 g 25g
11494967 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484967 Supplier Thermo Scientific Alfa Aesar Supplier No. B21899.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3586-14-9 | C13H12O | 184.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3586-14-9
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.24
MDL Number MFCD00008531
InChI Key UDONPJKEOAWFGI-UHFFFAOYSA-N
Synonym 3-phenoxytoluene, 3-methyldiphenyl ether, m-phenoxytoluene, phenyl m-tolyl ether, ether, phenyl m-tolyl, benzene, 1-methyl-3-phenoxy, m-methylphenyl phenyl ether, 3-methylphenyl phenyl ether, unii-2zzi1z67wr
PubChem CID 19165
IUPAC Name 1-methyl-3-phenoxybenzene
SMILES CC1=CC=CC(OC2=CC=CC=C2)=C1

Specifications

Density 1.052
Boiling Point 271°C to 273°C
Flash Point >110°C (230°F)
Refractive Index 1.574
Quantity 25 g
Beilstein 2045714
Formula Weight 184.24
Percent Purity 97%
Chemical Name or Material 3-Phenoxytoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.