missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxythiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10079341
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10079341 1 g 1g
10066464 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10079341 Supplier Thermo Scientific Maybridge Supplier No. KM05428DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 132706-25-3
Molecular Formula C11H8O2S
Molecular Weight (g/mol) 204.24
MDL Number MFCD00178748
InChI Key QHJDRZXMSDASSH-UHFFFAOYSA-N
Synonym 2-thiophenecarboxaldehyde,3-phenoxy, 3-phenoxythiophene-2-carboxaldehyde, acmc-1c0te, 3-phenoxy-2-thiophenecarbaldehyde, 3-phenoxythiophen-2-carboxaldehyde, 3-phenoxy-2-thiophenecarboxaldehyde
PubChem CID 2778987
IUPAC Name 3-phenoxythiophene-2-carbaldehyde
SMILES O=CC1=C(OC2=CC=CC=C2)C=CS1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 204.25
Percent Purity 97%
Chemical Name or Material 3-Phenoxythiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.