missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxypiperidine, 97%

Product Code. 11499620
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11499620

Brand: Thermo Scientific Alfa Aesar H50912.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 151666-08-9 | C11H15NO | 177.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151666-08-9
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD04117746
InChI Key QDPNIJKUTWDGMV-UHFFFAOYNA-N
Synonym piperidine, 3-phenoxy, 3-phenoxy-piperidine, piperidine,3-phenoxy, 3-phenoxypiperidine,, 3-phenoxy piperidine, acmc-20a6se
PubChem CID 3730581
IUPAC Name 3-phenoxypiperidine
SMILES C1CNCC(C1)OC1=CC=CC=C1

Specifications

Refractive Index 1.543
Quantity 250 mg
Formula Weight 177.25
Percent Purity 97%
Chemical Name or Material 3-Phenoxypiperidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.