missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxyphenylacetic acid, 98%

Product Code. 11333216
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333216 5 g 5g
11323216 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333216 Supplier Thermo Scientific Alfa Aesar Supplier No. L12524.06
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 32852-81-6 | C14H12O3 | 228.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32852-81-6
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00016826
InChI Key LEMRHTTWKDVQEI-UHFFFAOYSA-N
Synonym 3-phenoxyphenylacetic acid, 2-3-phenoxyphenyl acetic acid, 3-phenoxyphenyl acetic acid, 3-phenoxy-phenyl-acetic acid, benzeneacetic acid, 3-phenoxy, 3-phenoxyphenylaceticacid, m-phenoxyphenylacetic acid, acmc-20a35b, 3-phenoxybenzeneacetic acid, 3-phenoxyphenyl-acetic acid
PubChem CID 141749
IUPAC Name 2-(3-phenoxyphenyl)acetic acid
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)CC(=O)O

Specifications

Melting Point 88°C to 90°C
Quantity 5 g
Beilstein 2730540
Formula Weight 228.25
Percent Purity 98%
Chemical Name or Material 3-Phenoxyphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.