missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzyl Alcohol, TRC

Product Code. 30179520
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179520 25 g 25g
30179209 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179520 Supplier Toronto Research Chemicals Supplier No. TRCP22805025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-35-2
Molecular Formula C13 H12 O2
Molecular Weight (g/mol) 200.23
Synonym Benzenemethanol, 3-phenoxy-, Benzyl alcohol, m-phenoxy- (7CI,8CI), (3-Phenoxyphenyl)methanol, 1-Hydroxymethyl-3-phenoxybenzene, 3-(Hydroxymethyl)diphenyl ether, 3-Phenoxybenzenemethanol, 3-Phenoxybenzyl alcohol, m-Phenoxybenzyl alcohol, m-Phenoxybenzyl alcohol
IUPAC Name (3-phenoxyphenyl)methanol
SMILES OCc1cccc(Oc2ccccc2)c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C13H12O2/c14-10-11-5-4-8-13(9-11)15-12-6-2-1-3-7-12/h1-9,14H,10H2
Formula Weight 200.0837
Chemical Name or Material 3-Phenoxybenzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.