missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzyl Alcohol, Mikromol™

Product Code. 17984366
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17984366 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17984366 Supplier Mikromol Supplier No. MM3154.01
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Phenoxybenzyl Alcohol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Phenoxybenzyl alcohol
CAS 13826-35-2
Analyte or Component Names 3-Phenoxybenzyl alcohol
Shipping Condition Room Temperature
Molecular Formula C13 H12 O2
Certifications/Compliance ISO 9001
Synonym Benzenemethanol, 3-phenoxy-, Benzyl alcohol, m-phenoxy- (7CI,8CI), (3-Phenoxyphenyl)methanol, 1-Hydroxymethyl-3-phenoxybenzene, 3-(Hydroxymethyl)diphenyl ether, 3-Phenoxybenzenemethanol, 3-Phenoxybenzyl alcohol, m-Phenoxybenzyl alcohol, m-Phenoxybenzyl alcohol, Permethrin Impurity C
InChI Formula InChI=1S/C13H12O2/c14-10-11-5-4-8-13(9-11)15-12-6-2-1-3-7-12/h1-9,14H,10H2
SMILES OCc1cccc(Oc2ccccc2)c1
IUPAC Name (3-phenoxyphenyl)methanol
Molecular Weight (g/mol) 200.23
Quantity 100 mg
Formula Weight 200.0837 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.