missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzyl alcohol, 98%

Product Code. 10307510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307510 100 g Glass bottle 100g
10436042 5 g Glass bottle 5g
10792031 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10307510 Supplier Thermo Scientific Acros Supplier No. 183771000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13826-35-2 | C13H12O2 | 200.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-35-2
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD00004636
InChI Key KGANAERDZBAECK-UHFFFAOYSA-N
Synonym 3-phenoxybenzyl alcohol, 3-phenoxyphenyl methanol, 3-phenoxybenzylalcohol, m-phenoxybenzyl alcohol, 3-phenoxybenzenemethanol, benzenemethanol, 3-phenoxy, 3-hydroxymethyl diphenyl ether, 3-phenoxybenzylic alcohol, benzyl alcohol, m-phenoxy, 3-pboh
PubChem CID 26295
ChEBI CHEBI:62527
IUPAC Name (3-phenoxyphenyl)methanol
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless or White
Density 1.1456g/mL
Boiling Point 135.0°C to 140.0°C (1.0 mmHg)
Flash Point 309°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula C6H5OC6H4CH2OH
Refractive Index 1.5920 to 1.5940
Quantity 100 g
Specific Gravity 1.1456
Solubility Information Solubility in water: insoluble in water.
Formula Weight 200.24
Percent Purity 98%
Physical Form Liquid or Solid
Chemical Name or Material 3-Phenoxybenzyl alcohol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Phenoxybenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.