missing translation for 'onlineSavingsMsg'
Learn More

3-Pentyn-1-ol, 99%

Product Code. 10552411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10552411 5 g Glass bottle 5g
10093903 1 g Glass bottle 1g
10183970 25 g Glass bottle 25g
11934621 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10552411 Supplier Thermo Scientific Acros Supplier No. 197330050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10229-10-4 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10229-10-4
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00002956
InChI Key IDYNOORNKYEHHO-UHFFFAOYSA-N
Synonym 3-pentyn-1-ol, 3-pentynol, acmc-1bxad
PubChem CID 66295
IUPAC Name pent-3-yn-1-ol
SMILES CC#CCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 0.9100g/mL
Boiling Point 154.0°C to 157.0°C
Flash Point 54°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C≡CCH2CH2OH
Refractive Index 1.4550 to 1.4570
Quantity 5 g
Beilstein 01,IV,2227
Specific Gravity 0.91
Formula Weight 84.12
Percent Purity 99%
Chemical Name or Material 3-Pentyn-1-ol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Pentyn-1-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.