missing translation for 'onlineSavingsMsg'
Learn More

3-Pentyn-1-ol, 98%

Product Code. 15448809
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448809 1 g 1g
15458809 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448809 Supplier Thermo Scientific Alfa Aesar Supplier No. L02951.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10229-10-4 | C5H8O | 84.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10229-10-4
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00002956
InChI Key IDYNOORNKYEHHO-UHFFFAOYSA-N
Synonym 3-pentyn-1-ol, 3-pentynol, acmc-1bxad
PubChem CID 66295
IUPAC Name pent-3-yn-1-ol
SMILES CC#CCCO

Specifications

Density 0.912
Boiling Point 154°C to 157°C
Flash Point 54°C (129°F)
Odor Recognizable
Linear Formula CH3C≡CCH2CH2OH
Refractive Index 1.456
Quantity 1 g
UN Number UN1987
Beilstein 1735967
Formula Weight 84.12
Percent Purity 98%
Chemical Name or Material 3-Pentyn-1-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.