missing translation for 'onlineSavingsMsg'
Learn More

3-Pentyl formate, 94%, Thermo Scientific™

Product Code. 11468947
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468947 5 g 5g
11478947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468947 Supplier Thermo Scientific Alfa Aesar Supplier No. B20566.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58368-67-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00049220
InChI Key YMJOAYHERILFIM-UHFFFAOYSA-N
Synonym 3-pentyl formate, acmc-1b22e, formic acid 1-ethylpropyl ester
PubChem CID 13878880
IUPAC Name pentan-3-yl formate
SMILES CCC(CC)OC=O

Specifications

Density 0.873
Boiling Point 116°C to 118°C
Quantity 5 g
UN Number UN3272
Formula Weight 116.16
Percent Purity 94%
Chemical Name or Material 3-Pentyl formate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.