missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pentanone, ReagentPlus™, ≥99.0% (GC), Honeywell™
Reagent Grade,≥99.0% (GC)
£14.16 - £414.31
Chemical Identifiers
CAS | 96-22-0 |
---|---|
Molecular Formula | C5H10O |
Molecular Weight (g/mol) | 86.13 |
MDL Number | MFCD00009320 |
InChI Key | FDPIMTJIUBPUKL-UHFFFAOYSA-N |
Synonym | 3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone |
PubChem CID | 7288 |
ChEBI | CHEBI:67886 |
IUPAC Name | pentan-3-one |
SMILES | CCC(=O)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15621190
|
Honeywell Research Chemicals
127604-100ML |
100 mL | |||||||
15671190
|
Honeywell Research Chemicals
127604-500ML |
500 mL | |||||||
15641190
|
Honeywell Research Chemicals
127604-1L |
1 L | |||||||
15651190
|
Honeywell Research Chemicals
127604-2.5L |
2.5 L | |||||||
15661190
|
Honeywell Research Chemicals
127604-20L |
20 L | |||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
96-22-0 | |
86.13 | |
FDPIMTJIUBPUKL-UHFFFAOYSA-N | |
7288 | |
pentan-3-one |
C5H10O | |
MFCD00009320 | |
3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone | |
CHEBI:67886 | |
CCC(=O)CC |
Specifications
96-22-0 | |
Colorless | |
102°C | |
Glass Bottle | |
CH3CH2COCH2CH3 | |
MFCD00009320 | |
635749 | |
Slightly soluble in water | |
CCC(=O)CC | |
86.13 | |
CHEBI:67886 | |
≥99.0% (GC) | |
3-Pentanone |
-39°C | |
0.813g/mL at 25°C | |
7°C (44.6°F) | |
C5H10O | |
n20/D 1.392(lit.) | |
UN1156 | |
3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone | |
FDPIMTJIUBPUKL-UHFFFAOYSA-N | |
pentan-3-one | |
7288 | |
86.13g/mol | |
ReagentPlus™ |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
H225-H319-H335-H336
missing translation for 'dotInformation' : Class 3 - PG 2 - Diethyl ketone
missing translation for 'einecsNumber' : 202-490-3
missing translation for 'rtecsNumber' : SA8050000