missing translation for 'onlineSavingsMsg'
Learn More

3-Pentanone, 98%, pure

Product Code. p-179364 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051680 500 mL Glass bottle 500mL
10398893 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10051680 Supplier Thermo Scientific Acros Supplier No. 155815000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-22-0 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-22-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00009320
InChI Key FDPIMTJIUBPUKL-UHFFFAOYSA-N
Synonym 3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone
PubChem CID 7288
ChEBI CHEBI:67886
IUPAC Name pentan-3-one
SMILES CCC(=O)CC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -40.0°C
Color Colorless
Density 0.8150g/mL
Boiling Point 100.0°C to 102.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5COC2H5
Refractive Index 1.3910 to 1.3930
Quantity 500 mL
Beilstein 01, IV, 3279
Merck Index 15, 3137
Specific Gravity 0.815
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in ethanol and acetone,miscible with many organic solvents
Viscosity 0.47 mPa.s (20°C)
Formula Weight 86.13
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material 3-Pentanone, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Pentanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.