missing translation for 'onlineSavingsMsg'
Learn More

3-Pentanol, 98%

Product Code. 10477710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10477710 25 g Glass bottle 25g
10193270 100 g Glass bottle 100g
10735241 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10477710 Supplier Thermo Scientific Acros Supplier No. 129990250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 584-02-1 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-02-1
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004574
InChI Key AQIXEPGDORPWBJ-UHFFFAOYSA-N
Synonym 3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary
PubChem CID 11428
ChEBI CHEBI:77519
IUPAC Name pentan-3-ol
SMILES CCC(CC)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -75°C
Color Colorless
Density 0.8150g/mL
Boiling Point 114°C to 116°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(OH)C2H5
Refractive Index 1.4090 to 1.411
Quantity 25 g
Beilstein 01, 385
Merck Index 15, 7232
Specific Gravity 0.815
Solubility Information Solubility in water: 5.5g/100mL (30°C). Other solubilities: soluble in alcohol and ether
Formula Weight 88.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Pentanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Pentanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.