missing translation for 'onlineSavingsMsg'
Learn More

3-Pentanol, 98+%

Product Code. 11342346
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342346 25 g 25g
11352346 100 g 100g
11362346 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342346 Supplier Thermo Scientific Alfa Aesar Supplier No. L06065.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 584-02-1 | C5H12O | 88.15 g/mol

3-Pentanol is widely used as a flavoring agent. It is biogenic oxygenated volatile organic compound (BOVOC) and is used as a reagent in the synthesis of pure bromopentanes for infrared standards.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pentanol is widely used as a flavoring agent. It is biogenic oxygenated volatile organic compound (BOVOC) and is used as a reagent in the synthesis of pure bromopentanes for infrared standards.

Solubility
Soluble in acetone, benzene, ethanol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-02-1
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004574
InChI Key AQIXEPGDORPWBJ-UHFFFAOYSA-N
Synonym 3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary
PubChem CID 11428
ChEBI CHEBI:77519
IUPAC Name pentan-3-ol
SMILES CCC(CC)O

Specifications

Melting Point -8°C
Boiling Point 114°C to 115°C
Flash Point 33°C (91°F)
Odor Characteristic
Quantity 25 g
UN Number UN1105
Beilstein 1730964
Merck Index 14,7120
Formula Weight 88.15
Percent Purity ≥98%
Chemical Name or Material 3-Pentanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Pentanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Pentanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.