Learn More
TERUMO 3-Part Syringes
Product Code. 17102724
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
30 mL
Connection Type:
Centric Luer Slip
Concentric Luer Lock
Concentric Luer Slip
Eccentric Luer Tip
Unit Size:
Pack of 100
Pack of 25
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Connection Type Unit Size
17102724 5 mL Concentric Luer Slip Pack of 100
17142714 1 mL Concentric Luer Slip Pack of 100
17152714 3 mL Concentric Luer Lock Pack of 100
17162714 3 mL Centric Luer Slip Pack of 100
17705049 10 mL Eccentric Luer Tip Pack of 100
16207060 30 mL Concentric Luer Slip Pack of 25
16217060 30 mL Concentric Luer Lock Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17102724 Supplier TERUMO Supplier No. 8MDSS05SE
Quantity Price
1 £17.05 / Pack of 100
18 £15.30 / Pack of 100
 Restricted Item
In Stock
Edge
Add to basket
missing translation for 'validMainframeMsg'
Siliconisation for optimal plunger mobility
  • Every single detail is the result of Terumo's long time experience in this field which is appreciated by users throughout the world
  • Siliconisation for optimal plunger mobility
  • The integral gasket ensures a fi rm and distinct end-stop
  • Clear graduated scale allows accurate dosage
  • Transparent barrel for visualisation
  • Finger grips to support easy handling
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Material Polypropylene (Barrel), Polypropylene (Plunger)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Syringe
Disposable Single-use
Graduations 0.2 mL
Volume (Metric) 5 mL
For Use With (Equipment) Hypodermic Needle
Connection Type Concentric Luer Slip
Graduated Yes
Sterility Sterile