missing translation for 'onlineSavingsMsg'
Learn More

3-(p-Tolyl)propionic acid, 97%, Thermo Scientific™

Product Code. 10534944 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10534944 1 g Glass bottle 1g
10483264 5 g Glass bottle 5g
10544944 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10534944 Supplier Thermo Scientific Acros Supplier No. 427360010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1505-50-6
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00016561
InChI Key LDYGRLNSOKABMM-UHFFFAOYSA-N
Synonym 3-4-methylphenyl propionic acid, 3-p-tolyl propionic acid, 3-p-tolyl propanoic acid, benzenepropanoic acid, 4-methyl, 3-4-methylphenyl propanoic acid, 4-methylphenylpropionic acid, 3-p-tolylpropanoic acid, p-methylhydrocinnamic acid, 4-methyl phenyl propionic acid, 4-methylhydrocinnamic acid
PubChem CID 73927
IUPAC Name 3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(CCC(O)=O)C=C1

Specifications

CAS Min % 96
CAS Max % 100
Melting Point 115.0°C to 118.0°C
Color Orange or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula HOOC(CH2)2C6H4CH3
Quantity 1 g
Formula Weight 164.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-(p-Tolyl)propionic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-(p-Tolyl)propionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.