missing translation for 'onlineSavingsMsg'
Learn More

3’-p-Hydroxy Paclitaxel (1mg/mL in Dimethyl Sulfoxide), TRC

Product Code. 30484224
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484224 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30484224 Supplier Toronto Research Chemicals Supplier No. TRCKIT98641X1ML
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 132160-32-8
Molecular Formula C47 H51 N O15
Quantity 1 mL
Synonym Benzenepropanoic acid, β-(benzoylamino)-α,4-dihydroxy-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-(benzoylamino)-α,4-dihydroxy-, 6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-4-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4β,4aβ,6β,9α(αR*,βS*),11α,12α,12aα,12bα]]-, 3'-Para-hydroxypaclitaxel, 3'-(4-Hydroxyphenyl)paclitaxel, 3'-p-Hydroxypaclitaxel, 3'-p-Hydroxy paclitaxel, (2R,3S)-3-(Benzoylamino)-2-hydroxy-3-(4-hydroxyphenyl)propanoic acid (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester
InChI Formula InChI=1S/C47H51NO15/c1-24-31(61-43(57)36(53)35(27-17-19-30(51)20-18-27)48-41(55)28-13-9-7-10-14-28)22-47(58)40(62-42(56)29-15-11-8-12-16-29)38-45(6,32(52)21-33-46(38,23-59-33)63-26(3)50)39(54)37(60-25(2)49)34(24)44(47,4)5/h7-20,31-33,35-38,40,51-53,58H,21-23H2,1-6H3,(H,48,55)/t31-,32-,33+,35-,36+,37+,38+,40-,45+,46-,47+/m0/s1
SMILES CC(=O)O[C@H]1C(=O)[C@]2(C)[C@@H](O)C[C@H]3OC[C@@]3(OC(=O)C)[C@@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)c6ccccc6)c7ccc(O)cc7)C(=C1C5(C)C)C
Molecular Weight (g/mol) 869.91
Formula Weight 869.3259
Concentration or Composition (by Analyte or Components) 3'-p-Hydroxy Paclitaxel 1 mg/mL
Chemical Name or Material 3'-p-Hydroxy Paclitaxel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.