missing translation for 'onlineSavingsMsg'
Learn More

3-(p-Chlorophenyl)-3-hydroxyphthalimidine, TRC

Product Code. 30137059
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137059 500 mg 500mg
30132674 100 mg 100mg
17910075 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137059 Supplier Toronto Research Chemicals Supplier No. TRCC381075500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 956-92-3
Molecular Formula C14 H10 Cl N O2
Molecular Weight (g/mol) 259.69
Synonym 3-(4-Chlorophenyl)-2,3-dihydro-3-hydroxy-1H-isoindol-1-one, C 3/76, Chlorthalidone Impurity J
IUPAC Name 3-(4-chlorophenyl)-3-hydroxy-isoindolin-1-one
SMILES OC1(NC(=O)c2ccccc12)c3ccc(Cl)cc3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H10ClNO2/c15-10-7-5-9(6-8-10)14(18)12-4-2-1-3-11(12)13(17)16-14/h1-8,18H,(H,16,17)
Formula Weight 259.04
Chemical Name or Material 3-(p-Chlorophenyl)-3-hydroxyphthalimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.