missing translation for 'onlineSavingsMsg'
Learn More

3-Oxocyclopentanecarboxylic acid, 97%

Product Code. 15486878
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15486878 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15486878 Supplier Thermo Scientific Alfa Aesar Supplier No. H60414.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-78-2 | C6H8O3 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 98-78-2
Melting Point 59°C to 62°C
Molecular Formula C6H8O3
MDL Number MFCD01320173
Quantity 1 g
Synonym 3-oxocyclopentanecarboxylic acid, 3-oxo-cyclopentane-carboxylic acid, 3-oxo cyclopentane 1-carboxylic acid, 3-oxo-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-oxo, 3-carboxycyclopentanone, cyclopentanone-3-carboxylic acid, 3-oxo-cyclopentanecarboxylic acid, 3-ketocyclopentanecarboxylic acid, 3-cyclopentanone-1 r-carboxylic acid
InChI Key RDSNBKRWKBMPOP-UHFFFAOYNA-N
SMILES OC(=O)C1CCC(=O)C1
IUPAC Name 3-oxocyclopentane-1-carboxylic acid
Molecular Weight (g/mol) 128.13
PubChem CID 227798
ChEBI CHEBI:71213
Formula Weight 128.13
Percent Purity 97%
Chemical Name or Material 3-Oxocyclopentanecarboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.