missing translation for 'onlineSavingsMsg'
Learn More

3-Oxoandrost-4-en-17beta-yl Acetate (Testosterone Acetate), Mikromol™

Product Code. 18081900
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18081900 100 mg 100mg
18061500 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18081900 Supplier Mikromol Supplier No. MM0323.05
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Oxoandrost-4-en-17beta-yl Acetate (Testosterone Acetate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone Acetate
CAS 1045-69-8
Analyte or Component Names Testosterone Acetate
Shipping Condition Room Temperature
Molecular Formula C21 H30 O3
Certifications/Compliance ISO 17025
Synonym Testosterone Imp. E (EP), Testosterone Isocaproate Imp. B (EP), Testosterone Propionate Imp. A (EP), Testosterone Acetate, 3-Oxoandrost-4-en-17beta-yl Acetate, (17beta)-17-(Acetyloxy)androst-4-en-3-one, Testosterone acetate, 17-O-Acetyltestosterone, 17beta-Acetoxy-4-androsten-3-one, 17beta-Acetoxy-Delta4-androsten-3-one, 17beta-Acetoxyandrost-4-ene-3-one, 17beta-Acetoxytestosterone, 17beta-Hydroxyandrost-4-en-3-one acetate, 17beta-Hydroxyandrost-4-en-3-one-17beta-acetate, 3-Oxoandrost-4-en-17beta-yl acetate, 4-Androsten-17beta-ol-3-one 17-acetate, Aceto-sterandryl, Aceto-testoviron, Amolisin, Androst-4-en-17beta-ol-3-one acetate, Androtest A, Deposteron, Farmatest, NSC 523836, Perandrone A, SKF 5647, Testosterone 17-acetate, Testosterone Impurity E, Testosterone Isocaproate Impurity B, Testosterone Propionate Impurity A
InChI Formula InChI=1S/C21H30O3/c1-13(22)24-19-7-6-17-16-5-4-14-12-15(23)8-10-20(14,2)18(16)9-11-21(17,19)3/h12,16-19H,4-11H2,1-3H3/t16-,17-,18-,19-,20-,21-/m0/s1
SMILES CC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 330.46
Quantity 100 mg
Formula Weight 330.2195 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.