missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carboxylic Acid, TRC

Product Code. 30379999
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Product Code. Quantity
30379999 50 mg
30373697 100 mg
30382058 500 mg
3 options
This item is not returnable. View return policy

Product Code. 30379999

Brand: Toronto Research Chemicals TRCE58875850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134997-87-8
Formula Weight 193.0375
InChI Formula InChI=1S/C9H7NO4/c11-8-4-14-7-2-1-5(9(12)13)3-6(7)10-8/h1-3H,4H2,(H,10,11)(H,12,13)
IUPAC Name 3-oxo-4H-1,4-benzoxazine-6-carboxylic acid
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.16
SMILES OC(=O)c1ccc2OCC(=O)Nc2c1
Synonym 3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carboxylic Acid, 3-Hydroxy-2H-1,4-benzoxazine-6-carboxylic Acid, 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic Acid, 3-Oxo-3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylic Acid, 3-Oxo-4H-1,4-benzoxazine-6-carboxylic Acid
Chemical Name or Material 3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carboxylic Acid

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.