Learn More
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-sulfonyl chloride, 97%, Maybridge
Brand: Maybridge CC46803DE
Code : 31
Additional Details : CAS Number : 31794-45-3 Weight : 0.05500kg Transport : UN number : 3261 Chem class : 1a Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
3-Oxo-3, 4-dihydro-2H-1, 4-benzoxazine-6-sulfonyl chloride | |
96.5 | |
97% | |
C8H6ClNO4S | |
MFCD05664887 |
31794-45-3 | |
100.0 | |
97% | |
247.66 |
Specifications
3-Oxo-3, 4-dihydro-2H-1, 4-benzoxazine-6-sulfonyl chloride | |
96.5 | |
97% | |
C8H6ClNO4S | |
MFCD05664887 |
31794-45-3 | |
100.0 | |
97% | |
247.66 | |
5g |