missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, Thermo Scientific™

£100.00 - £275.00

Chemical Identifiers

CAS 943994-02-3
Molecular Formula C14H18BNO4
Molecular Weight (g/mol) 275.111
MDL Number MFCD12755796
InChI Key ZXOSNHPLTJAXSA-UHFFFAOYSA-N
Synonym 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 3-oxo-2h,4h-benzo b 1,4 oxazine-6-boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,4-dihydro-1,4-benzoxazin-3-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 3-oxo-3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-4h-1,4-benzoxazin-3-one
PubChem CID 46856474
IUPAC Name 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-1,4-benzoxazin-3-one
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)OCC(=O)N3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15407116
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H62168.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15497106
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H62168.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

943994-02-3
275.111
ZXOSNHPLTJAXSA-UHFFFAOYSA-N
46856474
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)OCC(=O)N3
C14H18BNO4
MFCD12755796
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 3-oxo-2h,4h-benzo b 1,4 oxazine-6-boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,4-dihydro-1,4-benzoxazin-3-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 3-oxo-3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-4h-1,4-benzoxazin-3-one
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-1,4-benzoxazin-3-one

Specifications

943994-02-3
MFCD12755796
ZXOSNHPLTJAXSA-UHFFFAOYSA-N
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-1,4-benzoxazin-3-one
46856474
Solid
C14H18BNO4
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 3-oxo-2h,4h-benzo b 1,4 oxazine-6-boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,4-dihydro-1,4-benzoxazin-3-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 3-oxo-3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-4h-1,4-benzoxazin-3-one
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)OCC(=O)N3
275.111
275.11
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications