missing translation for 'onlineSavingsMsg'
Learn More

3-O-Dinitrobenzoyl-25-hydroxy Vitamin D2, TRC

Product Code. 30381810
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381810 10 mg 10mg
30383329 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30381810 Supplier Toronto Research Chemicals Supplier No. TRCD47973010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-O-Dinitrobenzoyl-25-hydroxy Vitamin D2
Analyte or Component Names 3-O-Dinitrobenzoyl-25-hydroxy Vitamin D2
Shipping Condition Room Temperature
Molecular Formula C35 H46 N2 O7
Synonym (3beta,5Z,7E,22E)-3-O-Dinitrobenzoyl-9,10-secoergosta-5,7,10(19),22-tetraene-3,25-diol
InChI Formula InChI=1S/C35H46N2O7/c1-22-10-14-30(44-33(38)27-18-28(36(40)41)21-29(19-27)37(42)43)20-26(22)13-12-25-8-7-17-35(6)31(15-16-32(25)35)23(2)9-11-24(3)34(4,5)39/h9,11-13,18-19,21,23-24,30-32,39H,1,7-8,10,14-17,20H2,2-6H3/b11-9+,25-12+,26-13-/t23-,24+,30+,31-,32+,35-/m1/s1
Solution Type Neat
SMILES C[C@H](\C=C\[C@H](C)C(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@H](CCC3=C)OC(=O)c4cc(cc(c4)[N+](=O)[O-])[N+](=O)[O-])\CCC[C@]12C
IUPAC Name [(1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(E,2R,5S)-6-hydroxy-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexyl] 3,5-dinitrobenzoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 606.749
Quantity 10 mg
Formula Weight 606.331
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.