missing translation for 'onlineSavingsMsg'
Learn More

3-O-Benzyl-1,2:5,6-Di-O-isopropylidene-Alpha-D-glucofuranose, TRC

Product Code. 30178968
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30178968 1 g 1g
30178948 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30178968 Supplier Toronto Research Chemicals Supplier No. TRCB2760001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18685-18-2
Molecular Formula C19 H26 O6
Molecular Weight (g/mol) 350.41
Synonym α-D-Glucofuranose, 1,2:5,6-bis-O-(1-methylethylidene)-3-O-(phenylmethyl)-, D-Glucose, 3-O-benzyl-1,2:5,6-di-O-isopropylidene- (6CI), Glucofuranose, 3-O-benzyl-1,2:5,6-di-O-isopropylidene- (7CI), Glucofuranose, 3-O-benzyl-1,2:5,6-di-O-isopropylidene-, α-D- (8CI), 1,2:5,6-Bis-O-(1-methylethylidene)-3-O-(phenylmethyl)-α-D-glucofuranose, Furo[2,3-d]-1,3-dioxole, α-D-glucofuranose deriv., 3-O-Benzyl-1,2:5,6-di-O-isopropylidene-α-D-glucofuranose, NSC 56267, 3-O-Benzyl-1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose
IUPAC Name (3aR,5R,6S,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-6-phenylmethoxy-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxole
SMILES CC1(C)OC[C@@H](O1)[C@H]2O[C@@H]3OC(C)(C)O[C@@H]3[C@H]2OCc4ccccc4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H26O6/c1-18(2)21-11-13(23-18)14-15(20-10-12-8-6-5-7-9-12)16-17(22-14)25-19(3,4)24-16/h5-9,13-17H,10-11H2,1-4H3/t13-,14-,15+,16-,17-/m1/s1
Formula Weight 350.1729
Chemical Name or Material 3-O-Benzyl-1,2:5,6-Di-O-isopropylidene-a-D-glucofuranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.