missing translation for 'onlineSavingsMsg'
Learn More

3-O-Benzyl-1,2:5,6-di-O-isopropylidene-alpha-D-glucofuranose, 98%, Thermo Scientific™

Product Code. 15490607
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490607 500 mg 500mg
15470607 1 g 1g
15480607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490607 Supplier Thermo Scientific Alfa Aesar Supplier No. J66578.ME

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18685-18-2
Molecular Formula C19H26O6
Molecular Weight (g/mol) 350.411
MDL Number MFCD00269929
InChI Key ZHFVGOMEUGAIJX-QTPCEWDRSA-N
Synonym 3-o-benzyl-1,2:5,6-di-o-isopropylidene-, a-d-glucofuranose, 1,2:5,6-bis-o-1-methylethylidene-3-o-phenylmethyl-, 4-3ar,5r,6as-6-benzyloxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl-2,2-dimethyl-1,3-dioxolane
PubChem CID 45038272
IUPAC Name (3aR,5R,6aS)-5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-6-phenylmethoxy-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxole
SMILES CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)OCC4=CC=CC=C4)C

Specifications

Color Colorless to Yellow
Density 1.094
Boiling Point 140°C to 142°C
Refractive Index 1.4925
Quantity 500 mg
Optical Rotation −30° (c=1 in Ethanol)
Formula Weight 350.41
Percent Purity 98%
Physical Form Oil
Chemical Name or Material 3-O-Benzyl-1,2:5,6-di-O-isopropylidene-alpha-D-glucofuranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.