missing translation for 'onlineSavingsMsg'
Learn More

3-Nonen-2-one, 96%

Product Code. 11419386
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419386 10 g 10g
11429386 50 g 50g
11439386 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419386 Supplier Thermo Scientific Alfa Aesar Supplier No. A19084.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14309-57-0 | C9H16O | 140.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14309-57-0
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00010241
InChI Key HDKLIZDXVUCLHQ-BQYQJAHWSA-N
Synonym 3-nonen-2-one, trans-3-nonen-2-one, e-non-3-en-2-one, e-3-nonen-2-one, 3-nonen-2-one, 3e, 3e-3-nonen-2-one, 3e-non-3-en-2-one, unii-fr0k39of16, 3e-nonen-2-one, 3-nonen-2-one fhfi
PubChem CID 5317045
SMILES CCCCC\C=C\C(C)=O

Specifications

Density 0.848
Boiling Point 85°C (12 mmHg)
Flash Point 81°C (177°F)
Refractive Index 1.449
Quantity 10 g
Beilstein 1743558
IUPAC Name (E)-non-3-en-2-one
Formula Weight 140.23
Percent Purity 96%
Chemical Name or Material 3-Nonen-2-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.