missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrostyrene, 97%, stabilized

Product Code. 10444585 Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
missing translation for 'unitSize'
10g
2.5g
This item is not returnable. View return policy

Product Code. 10444585

missing translation for 'mfr': Thermo Scientific Acros 128990100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-39-0 | C8H7NO2 | 149.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-39-0
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00007276
InChI Key SYZVQXIUVGKCBJ-UHFFFAOYSA-N
Synonym 3-nitrostyrene, 1-nitro-3-vinylbenzene, m-nitrostyrene, benzene, 1-ethenyl-3-nitro, styrene, m-nitro, 1-nitro-3-vinyl-benzene, 3-nitrophenyl ethene, b-nitrostyrene, m-vinylnitrobenzene, 3-ethenylnitrobenzene
PubChem CID 68514
IUPAC Name 1-ethenyl-3-nitrobenzene
SMILES C=CC1=CC(=CC=C1)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -5°C
Color White
Density 1.0700g/mL
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC6H4NO2
Refractive Index 1.5820 to 1.584
Quantity 10 g
Beilstein 05, 478
Specific Gravity 1.07
Stabilizer approx. 0.1% 3, 5-di-tert-butylcatechol
Formula Weight 149.15
Percent Purity 97%
Physical Form Needles or Powder
Chemical Name or Material 3-Nitrostyrene
Show More Show Less
compliance-icons
Product Identifier
  • 3-Nitrostyrene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.