missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrosalicylic Acid, TRC

Product Code. 30011684
Change view
Click to view available options
Quantity:
100 mg
5 g
Unit Size:
100mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011684 5 g 5g
30006491 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011684 Supplier Toronto Research Chemicals Supplier No. TRCN5209005G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-38-1
Molecular Formula C7 H5 N O5
Molecular Weight (g/mol) 183.12
Synonym Salicylic acid, 3-nitro- (7CI,8CI), 2-Hydroxy-3-nitrobenzoic acid, 3-Nitrosalicylic acid, NSC 182
IUPAC Name 2-hydroxy-3-nitrobenzoic acid
SMILES OC(=O)c1cccc(c1O)[N+](=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H5NO5/c9-6-4(7(10)11)2-1-3-5(6)8(12)13/h1-3,9H,(H,10,11)
Formula Weight 183.0168
Percent Purity >95%
Chemical Name or Material 3-Nitrosalicylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.