missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrosalicylic acid, 98%

Product Code. 10431674 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10431674 5 g 5g
10112874 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10431674 Supplier Thermo Scientific Acros Supplier No. 399820050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85-38-1 | C7H5NO5 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-38-1
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
InChI Key WWWFHFGUOIQNJC-UHFFFAOYSA-N
Synonym 3-nitrosalicylic acid, 2-hydroxy-3-nitrobenzoic acid, benzoic acid, 2-hydroxy-3-nitro, salicylic acid, 3-nitro, 2-hydroxy-3-nitro-benzoic acid, 3-nitro-salicylsaure, 3-nitrosalicyclic acid, wln: wnr bq cvq, acmc-209q5q
PubChem CID 6807
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])O)C(=O)O

Specifications

Melting Point 142°C to 147°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula O2NC6H3(OH)COOH
Quantity 5 g
Merck Index 15,6717
Formula Weight 183.12
Percent Purity 98%
Chemical Name or Material 3-Nitrosalicylic acid
compliance-icons
Product Identifier
  • 3-Nitrosalicylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.